ADV465749740 AChemBlock

8-Chloro-1-methyl-6-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-3,4-dihydro-1H-quinolin-2-one

  • MDL number MFCD22415245
  • Cas Number : 1427588-44-0
  • Molecular Weight : 321.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/8-chloro-1-methyl-6-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-3-4-dihydro-1h-quinolin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8697
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart