ADV465749773 AChemBlock

KX1-004

  • MDL number MFCD25977063
  • Cas Number : 518058-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/kx1-004.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10217
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart