ADV465749779 AChemBlock

Iodomethyl)cyclobutane

  • MDL number MFCD06410737
  • Cas Number : 16408-62-1
  • Molecular Weight : 196.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/iodomethyl-cyclobutane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8778
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart