ADV465749817 AChemBlock

Potassium trifluoro(tetrahydro-2H-pyran-3-yl)borate

  • MDL number MFCD09992940
  • Cas Number : 1430219-80-9
  • Molecular Weight : 192.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/potassium-trifluoro-tetrahydro-2h-pyran-3-yl-borate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K10843
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart