ADV465749820 AChemBlock

Potassium {[(tert-butoxycarbonyl)amino]methyl} trifluoroborate

  • MDL number MFCD19686142
  • Cas Number : 1314538-55-0
  • Molecular Weight : 237.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/potassium-tert-butoxycarbonyl-amino-methyl-trifluoroborate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K10909
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart