ADV465749821 AChemBlock

Ethyl 2-cyclopropylthiazole-4-carboxylate

  • MDL number MFCD22988964
  • Cas Number : 135207-08-8
  • Molecular Weight : 197.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-cyclopropylthiazole-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K10910
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart