ADV465749822 AChemBlock

chlorodimethyl(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane

  • MDL number MFCD28401063
  • Cas Number : 929887-16-1
  • Molecular Weight : 220.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-3-2-dioxaborolane-2-chlorodimethylsilyl-4-4-5-5-tetramethyl.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K10911
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart