ADV465749838 AChemBlock

(2S)-FMOC-4,4-difluoro-pyrrolidine-2-carboxilic acid

  • MDL number MFCD01860709
  • Cas Number : 203866-21-1
  • Molecular Weight : 373.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2s-fmoc-4-4-difluoro-pyrrolidine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8836
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart