ADV465749841 AChemBlock

Tert-butyl 4-((methylsulfonyl)methyl)piperidine-1-carboxylate

  • MDL number MFCD19440828
  • Cas Number : 194872-09-8
  • Molecular Weight : 277.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-methylsulfonyl-methyl-piperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8849
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart