ADV465749844 AChemBlock

Methyl 4-((tert-butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylate

  • MDL number MFCD22577782
  • Cas Number : 943845-74-7
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-tert-butoxycarbonyl-amino-bicyclo-2-2-2-octane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8854
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart