ADV465749876 AChemBlock

2-Methylimidazo[1,2-a]pyridine-8-carboxylic acid

  • MDL number MFCD11886629
  • Cas Number : 133427-10-8
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methylimidazo-1-2-a-pyridine-8-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9889
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart