ADV465749878 AChemBlock

(3S)-N,1-dimethylpyrrolidin-3-amine

  • MDL number MFCD19220666
  • Cas Number : 792969-63-2
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-n-1-dimethylpyrrolidin-3-amine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9891
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart