ADV465749891 AChemBlock

4-Bromo-1,3-thiazole-2-carbonitrile

  • MDL number MFCD09607690
  • Cas Number : 1017781-52-0
  • Molecular Weight : 189.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-3-thiazole-2-carbonitrile.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9908
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart