ADV465749893 AChemBlock

tert-Butyl (3S)-3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]pyrrolidine-1-carboxylate

  • MDL number MFCD28501275
  • Cas Number : 1175273-55-8
  • Molecular Weight : 363.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-3-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-pyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9910
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart