ADV465749899 AChemBlock

tert-Butyl 3-oxo-2-oxa-8-azaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD24467002
  • Cas Number : 203662-19-5
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-oxo-2-oxa-8-azaspiro-4-5-decane-8-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9917
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart