ADV465749903 AChemBlock

Ethyl 2-{[(tert-butoxy)carbonyl]amino}cyclopropane-1-carboxylate

  • MDL number MFCD19689177
  • Cas Number : 613261-19-1
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-tert-butoxy-carbonyl-amino-cyclopropane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9921
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart