ADV465749904 AChemBlock

3-Phenoxy-cyclobutanecarboxylic acid

  • MDL number MFCD17014858
  • Cas Number : 1263284-46-3
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-phenoxy-cyclobutanecarboxyli-c-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9922
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart