ADV465749913 AChemBlock

(6-Chloropyridazin-3-yl)methanol

  • MDL number MFCD11977451
  • Cas Number : 1011487-94-7
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-chloropyridazin-3-yl-methanol.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9934
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart