ADV465749915 AChemBlock

(1R,4R)-2-Oxa-5-azabicyclo[2.2.1]heptane hemioxalate

  • MDL number MFCD27988025
  • Cas Number : 1523530-74-6
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-2-oxa-5-azabicyclo-2-2-1-heptane-hemioxalate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9936
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart