ADV465749922 AChemBlock

2-(5-Fluoropyridin-2-yl)acetic acid

  • MDL number MFCD09924935
  • Cas Number : 1000515-83-2
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-5-fluoropyridin-2-yl-acetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9943
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart