ADV465749923 AChemBlock

6-Benzyl 1-ethyl 6-azaspiro[2.5]octane-1,6-dicarboxylate

  • MDL number MFCD20265613
  • Cas Number : 147610-84-2
  • Molecular Weight : 317.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-benzyl-1-ethyl-6-azaspiro-2-5-octane-1-6-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9944
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart