ADV465749928 AChemBlock

(3-Methylpyridin-4-yl)boronic acid

  • MDL number MFCD03095365
  • Cas Number : 894808-72-1
  • Molecular Weight : 136.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-methylpyridin-4-yl-boronic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9950
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart