ADV465749932 AChemBlock

2-(Methoxymethyl)azetidine

  • MDL number MFCD23105901
  • Cas Number : 1290136-94-5
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methoxymethyl-azetidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9954
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart