ADV465749938 AChemBlock

4-Bromo-1,3-dimethyl-1H-pyrazol-5-amine

  • MDL number MFCD06805209
  • Cas Number : 19848-99-8
  • Molecular Weight : 190.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-3-dimethyl-1h-pyrazol-5-amine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9960
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart