ADV465749940 AChemBlock

7-Iodo-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD13183303
  • Cas Number : 1100318-98-6
  • Molecular Weight : 245.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/7-iodo-3h-imidazo-4-5-b-pyridine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9963
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart