ADV465749944 AChemBlock

6-Chloro-4-nitro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD09880160
  • Cas Number : 688781-87-5
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-nitro-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9967
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart