ADV465749955 AChemBlock

6-Chloro-4-methyl-3-nitropyridin-2-amine

  • MDL number MFCD17016063
  • Cas Number : 863878-22-2
  • Molecular Weight : 187.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-methyl-3-nitropyridin-2-amine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9978
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart