ADV465749964 AChemBlock

(2R,3S)-3-hydroxypyrrolidine-2-carboxylic acid; trifluoroacetic acid

  • MDL number MFCD28964051
  • Cas Number : 1818843-17-2
  • Molecular Weight : 245.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-3s-3-hydroxypyrrolidine-2-carboxylic-acid-trifluoroacetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9988
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart