ADV465749966 AChemBlock

2-(Azetidin-3-yl)acetonitrile hydrochloride

  • MDL number MFCD28501222
  • Cas Number : 1423057-36-6
  • Molecular Weight : 132.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-azetidin-3-yl-acetonitrile-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9990
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart