ADV465749967 AChemBlock

tert-Butyl 5'-bromo-3'H-spiro[azetidine-3,1'-[2]benzofuran]-1-carboxylate

  • MDL number MFCD22689962
  • Cas Number : 1398609-80-7
  • Molecular Weight : 340.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-bromo-3-h-spiro-azetidine-3-1-2-benzofuran-1-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9992
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart