ADV465749968 AChemBlock

6-Bromo-5-chloro-1H-indazole

  • MDL number MFCD22689251
  • Cas Number : 1305208-02-9
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Wheat powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-5-chloro-1h-indazole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9993
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart