ADV465749972 AChemBlock

(1R,3R,5R)-2-Boc-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

  • MDL number MFCD16621713
  • Cas Number : 1148048-39-8
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-5r-2-boc-2-azabicyclo-3-1-0-hexane-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9997
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart