ADV465749981 AChemBlock

2-Methyl-2h-indazole-3,7-dicarbaldehyde

  • MDL number MFCD28501561
  • Cas Number : 1788041-66-6
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-2h-indazole-3-7-dicarbaldehyde.html

Safety Information

signalword WARNING
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13007
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart