ADV465749982 AChemBlock

5'-Bromo-1'-ethenyl-1',2'-dihydrospiro[cyclopropane-1,3'-indole]-2'-one

  • MDL number MFCD29127302
  • Cas Number : 1788054-77-2
  • Molecular Weight : 264.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-ethenyl-1-2-dihydrospiro-cyclopropane-1-3-indole-2-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13008
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart