ADV465749985 AChemBlock

6-Methyl-7H-pyrrolo[2,3-d]pyrimidin-2-amine

  • MDL number MFCD27987418
  • Cas Number : 1638760-18-5
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-7h-pyrrolo-2-3-d-pyrimidin-2-amine.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13012
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart