ADV465749986 AChemBlock

Methyl 4-bromo-2-oxo-2,3-dihydro-1H-indole-6-carboxylate

  • MDL number MFCD27987646
  • Cas Number : 1638768-53-2
  • Molecular Weight : 270.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-2-oxo-2-3-dihydro-1h-indole-6-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13013
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart