ADV465749987 AChemBlock

(3-Methyl-1H-indol-5-yl)boronic acid

  • MDL number MFCD27987717
  • Cas Number : 590417-54-2
  • Molecular Weight : 175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1h-indol-5-yl-boronic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13014
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart