ADV465749988 AChemBlock

Bicyclo[2.2.1]heptane-1,4-diamine dihydrochloride

  • MDL number MFCD28501871
  • Cas Number : 1818847-46-9
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/bicyclo-2-2-1-heptane-1-4-diamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13015
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart