ADV465749993 AChemBlock

3-Nitro-4-[(oxan-4-ylmethyl)amino]benzene-1-sulfonamide

  • MDL number MFCD28142285
  • Cas Number : 1228779-96-1
  • Molecular Weight : 315.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-nitro-4-oxan-4-ylmethyl-amino-benzene-1-sulfonamide.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13020
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart