ADV465749994 AChemBlock

(S)-tert-butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide

  • MDL number MFCD09833247
  • Cas Number : 396074-50-3
  • Molecular Weight : 237.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-tert-butyl-5-methyl-1-2-3-oxathiazolidine-3-carboxylate-2-2-dioxide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13021
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart