ADV465749996 AChemBlock

5-Bromo-2-methyl-7-nitro-2H-indazole

  • MDL number MFCD21642023
  • Cas Number : 1363383-35-0
  • Molecular Weight : 256.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-methyl-7-nitro-2h-indazole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13023
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart