ADV465749999 AChemBlock

1-(2-Methyl-2H-indazol-7-yl)ethan-1-one

  • MDL number MFCD11869773
  • Cas Number : 1159511-30-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-2-methyl-2h-indazol-7-yl-ethan-1-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13026
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart