ADV465750012 AChemBlock

tert-Butyl N-[(4-methylpiperidin-4-yl)methyl]carbamate hydrochloride

  • MDL number MFCD23105887
  • Cas Number : 1523618-21-4
  • Molecular Weight : 264.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-methylpiperidin-4-yl-methyl-carbamate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13042
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart