ADV465750013 AChemBlock

2-Methyl-1H-pyrrolo[3,2-c]pyridine

  • MDL number MFCD12923617
  • Cas Number : 113975-37-4
  • Molecular Weight : 132.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-1h-pyrrolo-3-2-c-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13043
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart