ADV465750014 AChemBlock

tert-Butyl 4-oxo-6-azaspiro[2.5]octane-6-carboxylate

  • MDL number MFCD21607170
  • Cas Number : 1101840-74-7
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-oxo-6-azaspiro-2-5-octane-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13044
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart