ADV465750032 AChemBlock

tert-butyl (1-methyl-4-oxocyclohexyl)carbamate

  • MDL number MFCD18374771
  • Cas Number : 412293-43-7
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/carbamic-acid-n-1-methyl-4-oxocyclohexyl-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13065
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart