ADV465750043 AChemBlock

3,3-Dimethyldihydro-2h-pyran-4(3h)-one

  • MDL number MFCD20484202
  • Cas Number : 625099-31-2
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow oil
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethyldihydro-2h-pyran-4-3h-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13076
pictograms GHS02
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart