ADV465750050 AChemBlock

Methyl 5-bromo-8-hydroxy-1,6-naphthyridine-7-carboxylate

  • MDL number MFCD13151938
  • Cas Number : 410544-37-5
  • Molecular Weight : 283.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-8-hydroxy-1-6-naphthyridine-7-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13085
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart