ADV465750051 AChemBlock

2-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13189359
  • Cas Number : 909562-81-8
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-methyl-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13086
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart