ADV465750062 AChemBlock

4,4,5,5-Tetrachlorospiro[2.2]pentane-1-carboxylic acid

  • MDL number MFCD28501578
  • Cas Number : 1788041-68-8
  • Molecular Weight : 249.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-4-5-5-tetrachlorospiro-2-2-pentane-1-carboxylic-acid.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13098
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart